Learn More
Medchemexpress LLC 4'-acetyl-N-[4-[4-(2-methoxyphenyl)-1-piperazinyl]butyl]-[1,1'-biphenyl]-4-carboxamide | 162408-66-4 | 99.9% | 485.62 g/mol | C30H35N3O3 | 50 MG
SDP
Catalog No. 5000213223
Encompass_Preferred
GR 103691 is a potent, selective dopamine D3 receptor antagonist used as a research reagent in pharmacology and neuroscience studies. It exhibits sub-nanomolar affinity for the D3 receptor and strong selectivity versus other dopamine receptor subtypes.
  • Potent D3 affinity (Ki ≈ 0.32-0.4 nM).
  • >100-fold selectivity over D1, D2, and D4 receptor subtypes.
  • High chemical purity (~99.9%) suitable for biochemical assays.
  • Well characterized molecular properties (molecular weight ≈ 485.6; formula C30H35N3O3).
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-13223 Supplier Medchemexpress LLC Supplier No. HY10138250MG
GR 103691 is a potent, selective dopamine D3 receptor antagonist used as a research reagent in pharmacology and neuroscience studies. It exhibits sub-nanomolar affinity for the D3 receptor and strong selectivity versus other dopamine receptor subtypes.
  • Potent D3 affinity (Ki ≈ 0.32-0.4 nM).
  • >100-fold selectivity over D1, D2, and D4 receptor subtypes.
  • High chemical purity (~99.9%) suitable for biochemical assays.
  • Well characterized molecular properties (molecular weight ≈ 485.6; formula C30H35N3O3).
  • Available in small research pack sizes for laboratory use.