Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-acetylamino-3-methyl-benzenesulfonyl chloride

Catalog No. CDS009227 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS009227 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS009227 Supplier Sigma Aldrich Supplier No. CDS009227100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N-ethylacetamide is a suitable nylon model for mechanistic studies of dye fading.n-ethylacetamide was used to investigate propylene carbonate and a mixture of n-methylformamide and n-ethylacetamide by broadband dielectric and mechanical shear spectroscopy
  • It was used in determination of reaction products of oh-oxidation of n-methylpyrrolidone under atmospheric conditions

Specifications

CAS 14988-21-7
Molecular Formula C9H10ClNO3S
Linear Formula C9H10ClNO3S
MDL Number MFCD06797216
Quantity 25 g
Molecular Weight (g/mol) 247.7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.