Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetylaminobenzaldehyde thiosemicarbazone, 98%, Thermo Scientific™

Catalog No. AC336520250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC336520250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC336520250 Supplier Thermo Scientific Chemicals Supplier No. 336520250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-06-3
Molecular Formula C10H12N4OS
Molecular Weight (g/mol) 236.293
MDL Number MFCD00022157
InChI Key SRVJKTDHMYAMHA-WUXMJOGZSA-N
Synonym thioacetazone, thiacetazone, amithiozone, ambathizon, benzothiozane, conteben, benzothiozon, benthiozone, parazone, thioacetazon
PubChem CID 9568512
IUPAC Name N-[4-[(E)-(carbamothioylhydrazinylidene)methyl]phenyl]acetamide
SMILES CC(=O)NC1=CC=C(C=C1)C=NNC(=S)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 25 g
Merck Index 15, 9442
Formula Weight 236.29
Chemical Name or Material 4-Acetylaminobenzaldehyde thiosemicarbazone, 98%
EINECSNumber 203-170-6
RTECSNumber AE3850000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.