Learn More
Medchemexpress LLC 4-Acetylbiphenyl | 92-91-1 | MFCD00008749 | 99.29% | 196.24 | 500 G
SDP
Catalog No. 5000453933
Encompass_Preferred
4-Acetylbiphenyl (ACBP) is a biphenyl compound. It can be utilized as a probe molecule to study the structure and function of proteins and to investigate the properties of lipid membranes.
  • Can be used as a probe molecule.
  • Useful for studying protein structure and function.
  • Applicable for researching lipid membrane properties.
Catalog No. 50-004-53933 Supplier Medchemexpress LLC Supplier No. HYW009877500G
Add to Cart
Add to Cart
4-Acetylbiphenyl (ACBP) is a biphenyl compound. It can be utilized as a probe molecule to study the structure and function of proteins and to investigate the properties of lipid membranes.
  • Can be used as a probe molecule.
  • Useful for studying protein structure and function.
  • Applicable for researching lipid membrane properties.