Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Acetylbutyric acid

Catalog No. A132045G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A132045G 5 g
A1320425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A132045G Supplier Sigma Aldrich Supplier No. A132045G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Acetylbutyric acid

Specifications

Melting Point 13°C to 14°C (lit.)
Density 1.09 g/mL (at 25°C (literature))
Boiling Point 274°C to 275°C (lit.)
Molecular Formula C6H10O3
Refractive Index n20/D 1.4451 (literature)
Linear Formula CH3CO(CH2)3 CO2H
MDL Number MFCD00004412
Quantity 5 g
Synonym 5-Ketohexanoic acid
Molecular Weight (g/mol) 130.14
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.