Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetylindan 98.0+%, TCI America™
SDP

Catalog No. A22961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22961G 1 g
A22965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22961G Supplier TCI America Supplier No. A22961G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38997-97-6
Molecular Formula C11H12O
Molecular Weight (g/mol) 160.216
MDL Number MFCD09033715
InChI Key FPHUGFGBPNVMQR-UHFFFAOYSA-N
PubChem CID 13657961
IUPAC Name 1-(2,3-dihydro-1H-inden-4-yl)ethanone
SMILES CC(=O)C1=C2CCCC2=CC=C1

Specifications

Melting Point 41°C
Color White-Yellow
Boiling Point 103°C
Quantity 1 g
Formula Weight 160.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Acetylindan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.