Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. A19075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1907-5G 5 g
A19071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1907-5G Supplier TCI America Supplier No. A19075G
Add to Cart
Add to Cart

Specifications

CAS 149104-90-5
Melting Point 239°C
Color White-Yellow
Molecular Formula C8H9BO3
Quantity 5 g
Synonym 4-acetylphenyl boronic acid, p-acetylphenylboronic acid, 4-acetylbenzeneboronic acid, 1-4-dihydroxyboranyl phenyl ethan-1-one, boronic acid, 4-acetylphenyl
InChI Key OBQRODBYVNIZJU-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)C(=O)C)(O)O
IUPAC Name (4-acetylphenyl)boronic acid
Molecular Weight (g/mol) 163.967
PubChem CID 3702122
Formula Weight 163.97
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.