Learn More
Medchemexpress LLC 4-allyltoluene | 3333-13-9 | MFCD00048186 | 97.0% | 132.20 g/mol | C10H12 | 250 MG
SDP
Catalog No. 5000208920
Encompass_Preferred
4-Allyltoluene is an aromatic small molecule used in insect olfaction research; it elicits antennal olfactory responses in Mediterranean fruit fly measured by electroantennography (EAG). Supplied at analytical purity for laboratory research, the compound is suitable for use in behavioral and electrophysiological assays.
  • Aromatic compound used in olfactory assays.
  • Elicits antennal response in Mediterranean fruit fly measured by EAG.
  • Analytical purity of 97.0%.
  • Molecular formula C10H12 and molecular weight 132.20 g/mol.
  • Available as 250 mg quantity for research applications.
Catalog No. 50-002-08920 Supplier Medchemexpress LLC Supplier No. 250MGHYW074514
4-Allyltoluene is an aromatic small molecule used in insect olfaction research; it elicits antennal olfactory responses in Mediterranean fruit fly measured by electroantennography (EAG). Supplied at analytical purity for laboratory research, the compound is suitable for use in behavioral and electrophysiological assays.
  • Aromatic compound used in olfactory assays.
  • Elicits antennal response in Mediterranean fruit fly measured by EAG.
  • Analytical purity of 97.0%.
  • Molecular formula C10H12 and molecular weight 132.20 g/mol.
  • Available as 250 mg quantity for research applications.