Learn More
Cayman Chemical 4-Allylcatechol, Purity: ≥98%, CAS: 1126-61-0, 100 mg
SDP
Catalog No. 502913769
Encompass_Preferred
A phenol with diverse biological activities; scavenges DPPH radicals, reduces the formation of TBARS, and inhibits xanthine oxidase in cell-free assays (IC50s = 20.17, 2, and 16.7 µM, respectively); cytotoxic to HT-29 cells (IC50 = 30 µg/ml); reduces paw edema and arthritic paw tissue homogenate levels of IL-1β, PGE2, LTB4, and NO in a mouse model of steam-killed M. tuberculosis-induced arthritis at 4 mg/kg; has been used as a precursor in the synthesis of, and is a metabolite of, safrole
Catalog No. 50-291-3769 Supplier Cayman Chemical Supplier No. 36542100 MG
A phenol with diverse biological activities; scavenges DPPH radicals, reduces the formation of TBARS, and inhibits xanthine oxidase in cell-free assays (IC50s = 20.17, 2, and 16.7 µM, respectively); cytotoxic to HT-29 cells (IC50 = 30 µg/ml); reduces paw edema and arthritic paw tissue homogenate levels of IL-1β, PGE2, LTB4, and NO in a mouse model of steam-killed M. tuberculosis-induced arthritis at 4 mg/kg; has been used as a precursor in the synthesis of, and is a metabolite of, safrole