Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-amino-1-(2-deoxy-2-fluoro-beta-D-arabinofuranosyl)pyrimidin-2(1H)-one | 56632-83-8 | MFCD07374399 | 99.9% | 245.21 | C9H12FN3O4 | 10 MG
SDP

Catalog No. 5000282611
Encompass_Preferred

2'-Deoxy-2'-fluoro-b-D-arabinocytidine is a fluorinated nucleoside analogue used in research on nucleoside metabolism and antitumor activity. It is supplied as a solid and as prepared DMSO solutions, and is characterized by high purity and defined storage recommendations for solids and stock solutions.

  • Fluorinated nucleoside analog suitable for biochemical and cancer research.
  • High purity: 99.92%.
  • Molecular formula C9H12FN3O4 and molecular weight 245.21.
  • Available as solid and as DMSO stock solution formats.
  • Solid is stable at room temperature; solutions should be stored at -80°C for long term.
  • Appearance: solid, facilitating handling and formulation.

Catalog No. 50-002-82611 Supplier Medchemexpress LLC Supplier No. HYW12878810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxy-2'-fluoro-b-D-arabinocytidine is a fluorinated nucleoside analogue used in research on nucleoside metabolism and antitumor activity. It is supplied as a solid and as prepared DMSO solutions, and is characterized by high purity and defined storage recommendations for solids and stock solutions.

  • Fluorinated nucleoside analog suitable for biochemical and cancer research.
  • High purity: 99.92%.
  • Molecular formula C9H12FN3O4 and molecular weight 245.21.
  • Available as solid and as DMSO stock solution formats.
  • Solid is stable at room temperature; solutions should be stored at -80°C for long term.
  • Appearance: solid, facilitating handling and formulation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.