Learn More
Enzo Life Sciences 4-Amino-1,8-naphthalimide (20mg). CAS: 1742-95-6
SDP
Supplier:  Enzo Life Sciences BMLAP1010020
Encompass_Preferred
Potent inhibitor of poly (ADP-ribose) polymerase-1 (PARP-1) (IC50=0.18μM). Reduces ischemia-reperfusion injury in the heart and skeletal muscle. Alternative name: 4-ANI. Purity: ≥96% (NMR). Solubility: Soluble in DMSO (5mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0305