Learn More
Cayman Chemical 4-amino-1,8-Naphthalimide, Purity: ≥95%, CAS: 1742-95-6, 50 mg
SDP
Catalog No. 502965208
Encompass_Preferred
An inhibitor of PARP (IC50 = 180 nM); blocks radiation-induced PARP in cancer cells, potentiating the cytotoxicity of γ-radiation, although it is not cytotoxic in the absence of radiation
Catalog No. 50-296-5208 Supplier Cayman Chemical Supplier No. 1850950 MG
An inhibitor of PARP (IC50 = 180 nM); blocks radiation-induced PARP in cancer cells, potentiating the cytotoxicity of γ-radiation, although it is not cytotoxic in the absence of radiation