Learn More
Accela Chembio Inc 4-amino-1-benzylpiperidine-4-carboxylic Acid | 25g | 39143-25-4 | MFCD00794952 | 95+% | Shelf Life: 900 Days | Light Sensitive/n2/+4
Supplier:  Accela Chembio Inc SY00403825G
Encompass_Preferred
4-amino-1-benzylpiperidine-4-carboxylic Acid | 25g | 39143-25-4 | MFCD00794952 | 95+% | Shelf Life: 900 Days | Light Sensitive/n2/+4
Catalog No. 50-353-952