Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Amino-1-benzylpiperidine

Catalog No. 19581225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-581-225G 25 g
19-581-25G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-581-225G Supplier Sigma Aldrich Supplier No. 19581225G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for synthesis of: Highly selective inhibitors of p38a mitogen-activated protein kinase Antiplasmodial compounds Dual activity cholinesterase and Aα-aggregation inhibitors Muscarinic acetylcholine receptor antagonist and beta 2 adrenoceptor agonists Photostable near-infrared cyanine dyes 99mTc-labeled piperidine analogues for targeting sigma receptors

Specifications

CAS 50541-93-0
Density 0.933 g/mL (at 25°C (literature))
Molecular Formula C12H18N2
Refractive Index n20/D 1.543 (literature)
Linear Formula C12H18N2
MDL Number MFCD00006504
Quantity 25 g
Synonym 1-(Phenylmethyl)-4-piperidinamine
Molecular Weight (g/mol) 190.28
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.