Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-1-methyl-3-propylpyrazole-5-carboxamide 98.0+%, TCI America™
SDP

Catalog No. A28575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28575G 5 g
A285725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28575G Supplier TCI America Supplier No. A28575G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 139756-02-8
Molecular Formula C8H14N4O
Molecular Weight (g/mol) 182.227
MDL Number MFCD02927682
InChI Key PZMXDLWWQHYXGY-UHFFFAOYSA-N
PubChem CID 3338044
ChEBI CHEBI:59006
IUPAC Name 4-amino-2-methyl-5-propylpyrazole-3-carboxamide
SMILES CCCC1=NN(C(=C1N)C(=O)N)C

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 5 g
Formula Weight 182.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-1-methyl-3-propylpyrazole-5-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.