Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-1-methylpiperidine 98.0+%, TCI America™
SDP

Catalog No. A204625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A204625G 25 g
A20465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A204625G Supplier TCI America Supplier No. A204625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 41838-46-4
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD01570549
InChI Key ALOCUZOKRULSAA-UHFFFAOYSA-N
PubChem CID 2737531
IUPAC Name 1-methylpiperidin-4-amine
SMILES CN1CCC(CC1)N

Specifications

Color Yellow
Boiling Point 83°C
Quantity 25 g
UN Number 2920
Formula Weight 114.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Amino-1-methylpiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.