Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-1-naphthalenecarbonitrile 98%, Thermo Scientific™

Catalog No. AC153510010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC153510010 1 g
AC153510050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC153510010 Supplier Thermo Scientific Chemicals Supplier No. 153510010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58728-64-6
Molecular Formula C11H8N2
Molecular Weight (g/mol) 168.199
MDL Number MFCD00004022
InChI Key LUSIZUFVMKYWGX-UHFFFAOYSA-N
Synonym 4-amino-1-naphthalenecarbonitrile, 4-amino-1-naphthonitrile, 1-naphthalenecarbonitrile, 4-amino, 4-cyano-1-naphthylamine, 1-amino-4-cyanonaphthalene, 1-naphthalenecarbonitrile,4-amino, acmc-1b1wx, 1-amino-4-cyano naphthalene, 4-amino-1-naphthonitrile #, 4-aminonaphthalenecarbonitrile
PubChem CID 42828
IUPAC Name 4-aminonaphthalene-1-carbonitrile
SMILES C1=CC=C2C(=C1)C(=CC=C2N)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula H2NC10H6CN
Quantity 1 g
Beilstein 14, 533
Formula Weight 168.2
Percent Purity ≥97.5%
Physical Form Fluffy Powder or Fine Needles
Chemical Name or Material 4-Amino-1-naphthalenecarbonitrile, 98%
Show More Show Less
EINECSNumber 261-412-6
RTECSNumber QJ1865000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.