Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-1-naphthol hydrochloride, 90%, technical

Catalog No. p-58255
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC158510050 5 g
AC158510250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC158510050 Supplier Thermo Scientific Chemicals Supplier No. 158510050
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 5959-56-8 | C10H9NO·HCl | 195.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5959-56-8
Molecular Formula C10H9NO·HCl
Molecular Weight (g/mol) 195.65
MDL Number MFCD00012562
InChI Key FDBQTRARWCKEJY-UHFFFAOYSA-N
Synonym 4-amino-1-naphthol hydrochloride, 4-aminonaphthalen-1-ol hydrochloride, 1-amino-4-naphthol hydrochloride, 1-naphthalenol, 4-amino-, hydrochloride, 4-amino-1-naphtholhydrochloride, 4-aminonaphthol, chloride, pcmd-cc-pab-253, 1-naphthalenol, 4-amino-, hydrochloride 1:1, acmc-1aogt, 4-amino-1-naphthol . hcl
PubChem CID 2723858
IUPAC Name 4-aminonaphthalen-1-ol;hydrochloride
SMILES C1=CC=C2C(=C1)C(=CC=C2O)N.Cl

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 273.0°C
Color Gray to Pink
Infrared Spectrum Authentic
Assay Percent Range 88% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NC10H6OH·HCl
Quantity 5 g
Beilstein 13, 667
Merck Index 10, 457
Formula Weight 195.65
Percent Purity 90%
Grade Technical
Physical Form Powder
Chemical Name or Material 4-Amino-1-naphthol hydrochloride, tech., 90%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 227-727-8
RTECSNumber QL3342000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.