Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-1-phenylpyrazolo[3,4-d]pyrimidine 98.0+%, TCI America™
SDP

Catalog No. A27371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27371G 1 g
A2737200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27371G Supplier TCI America Supplier No. A27371G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5334-30-5
Molecular Formula C11H9N5
Molecular Weight (g/mol) 211.23
MDL Number MFCD00022901
InChI Key KKDPIZPUTYIBFX-UHFFFAOYSA-N
Synonym 4-amino-1-phenylpyrazolo 3,4-d pyrimidine, 1-phenyl-1h-pyrazolo 3,4-d pyrimidin-4-amine, pp3, pp 3, 1-phenylpyrazolo 3,4-d pyrimidin-4-amine, chembl65063, 1-phenyl-1h-pyrazolo 3,4-d pyrimidin-4-ylamine, 4-amino-7-phenylpyrazol 3,4-d pyrimidine, 4-amino-1-phenylpyrazolo 3,4-e pyrimidine, 1h-pyrazolo 3,4-d pyrimidin-4-amine, 1-phenyl
PubChem CID 4879
IUPAC Name 1-phenyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
SMILES NC1=C2C=NN(C2=NC=N1)C1=CC=CC=C1

Specifications

Melting Point 216°C
Color White-Yellow
Quantity 1 g
Formula Weight 211.23
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-1-phenylpyrazolo[3,4-d]pyrimidine
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.