Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-1H-pyrazole, 95%

Catalog No. AAH28921MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH28921MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH28921MC Supplier Thermo Scientific Chemicals Supplier No. H28921MC
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 28466-26-4 | C3H5N3 | 83.094 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28466-26-4
Molecular Formula C3H5N3
Molecular Weight (g/mol) 83.094
MDL Number MFCD01693729
InChI Key AXINVSXSGNSVLV-UHFFFAOYSA-N
Synonym 4-aminopyrazole, 4-amino-1h-pyrazole, 1h-pyrazol-4-ylamine, 9ci, pyrazole, 4-amino, 1h-pyrazole-4-amine, pyrazole-4-ylamine, 4-nh2-pyrazole, 1-h-pyrazol-4-amine, pubchem14988
PubChem CID 78035
IUPAC Name 1H-pyrazol-4-amine
SMILES C1=C(C=NN1)N

Specifications

Melting Point 72°C to 77°C
Quantity 100 mg
Sensitivity Hygroscopic
Formula Weight 83.09
Percent Purity 95%
Chemical Name or Material 4-Amino-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber UQ4993000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.