Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2,2,5,5-tetramethyl-3-imidazoline-1-yloxy, 99%, free radical, Thermo Scientific™

Catalog No. AC290351000
Change view
Click to view available options
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC290351000 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC290351000 Supplier Thermo Scientific Chemicals Supplier No. 290351000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Polymerization reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 69826-42-2
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C7H14N3O
Quantity 100mg
Synonym 4-amino-2,2,5,5-tetramethyl-3-imidazoli-ne-1-yloxyl free radical, 4-amino-2.5-dihydro-2,2,5,5-tetramethyl-1h-imidazol-1-yl oxy
InChI Key JOUQHALJTYAVRQ-UHFFFAOYSA-N
SMILES CC1(C(=NC(N1[O])(C)C)N)C
IUPAC Name 1-$l^{1}-oxidanyl-2,2,5,5-tetramethylimidazol-4-amine
Molecular Weight (g/mol) 156.209
PubChem CID 2724894
Formula Weight 156.21
Percent Purity ≥98.5%
Chemical Name or Material 4-Amino-2,2,5,5-tetramethyl-3-imidazoline-1-yloxy, 99%
Show More Show Less
Recommended Storage Indoors +20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.