Learn More
Medchemexpress LLC 4-amino-2-[4-[1-(benzyloxycarbonyl)-2(S)-[(1,1-dimethylethyl)amino]carbonyl-piperazinyl]-6,7-dimethoxyqu | 189349-50-6 | 99.9% | 522.60 | C27H34N6O5 | 5 MG
SDP
Catalog No. 5000212908
Encompass_Preferred
L-765314 is a research reagent: a selective α1B (alpha-1B) adrenergic receptor antagonist used to probe α1B-mediated signaling in pharmacology and neuroscience studies. It is supplied as a solid powder with documented affinity and high reported purity, suitable for in vitro and in vivo experiments.
  • High affinity for α1B receptors (human Ki 2.0 nM; rat Ki 5.4 nM).
  • High purity suitable for research (reported 99.92%).
  • Solid powder form enabling flexible formulation and dosing.
  • Molecular weight 522.60 and formula C27H34N6O5 for precise calculations.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-12908 Supplier Medchemexpress LLC Supplier No. HY1013855MG
L-765314 is a research reagent: a selective α1B (alpha-1B) adrenergic receptor antagonist used to probe α1B-mediated signaling in pharmacology and neuroscience studies. It is supplied as a solid powder with documented affinity and high reported purity, suitable for in vitro and in vivo experiments.
  • High affinity for α1B receptors (human Ki 2.0 nM; rat Ki 5.4 nM).
  • High purity suitable for research (reported 99.92%).
  • Solid powder form enabling flexible formulation and dosing.
  • Molecular weight 522.60 and formula C27H34N6O5 for precise calculations.
  • Stable under recommended storage conditions for long-term use.