Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Amino-2,6-dichlorophenol

Catalog No. A4840725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A4840725G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A4840725G Supplier Sigma Aldrich Supplier No. A4840725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Traditionally challenging to access, this hindered amine provides a chemically differentiated building block for organic synthesis and medicinal chemistry, specifically for preparation of drug candidates containing hindered amine motifs
  • Additionally, this and over 80 other sterically hindered secondary amines in the sigma-aldrich portfolio provide structurally distinct screening compounds
  • Screen several through aldrich market select
  • This hindered amine was accessed via a recent publication reported by baran and coworkers using an innovative hydroamination method.product of kemxtree

Specifications

CAS 5930-28-9
Melting Point 167°C to 170°C (lit.)
Molecular Formula C6H5Cl2NO
Linear Formula H2NC6H2(Cl)2 OH
MDL Number MFCD00007875
Quantity 250 mg
Molecular Weight (g/mol) 178.02
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.