Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-amino-2,6-dichloropyrimidine | 10132-07-7 | MFCD00038015 | >98.0% | 163.99 g/mol | C4H3Cl2N3 | 25 G
SDP

Catalog No. 5000302803
Encompass_Preferred

4-Amino-2,6-dichloropyrimidine (CAS 10132-07-7) is an organic intermediate and biochemical reagent used in synthetic chemistry and life-science research. It serves as a versatile building block for heterocyclic synthesis and for development of pharmaceutical and agrochemical compounds.

  • High purity (>98.0%) suitable for research and synthesis.
  • Molecular formula C4H3Cl2N3 and molecular weight 163.99 g/mol.
  • White to yellow/orange powder or crystals.
  • Useful as a building block in heterocyclic and medicinal chemistry.
  • Store in a cool, dry place, typically 2-8 °C where recommended.

Catalog No. 50-003-02803 Supplier Medchemexpress LLC Supplier No. 25GHYW018744
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Amino-2,6-dichloropyrimidine (CAS 10132-07-7) is an organic intermediate and biochemical reagent used in synthetic chemistry and life-science research. It serves as a versatile building block for heterocyclic synthesis and for development of pharmaceutical and agrochemical compounds.

  • High purity (>98.0%) suitable for research and synthesis.
  • Molecular formula C4H3Cl2N3 and molecular weight 163.99 g/mol.
  • White to yellow/orange powder or crystals.
  • Useful as a building block in heterocyclic and medicinal chemistry.
  • Store in a cool, dry place, typically 2-8 °C where recommended.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.