Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-chloro-α-(4-chlorophenyl)-5-methylbenzeneacetonitrile 97%, Thermo Scientific™

Catalog No. AC133370500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC133370500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC133370500 Supplier Thermo Scientific Chemicals Supplier No. 133370500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61437-85-2
Molecular Formula C15H12Cl2N2
Molecular Weight (g/mol) 291.175
InChI Key SCNVPMWFNDBBQS-UHFFFAOYSA-N
Synonym 2-4-amino-2-chloro-5-methylphenyl-2-4-chlorophenyl acetonitrile, 2-amino-4-chloro-5-4 inverted exclamation mark-chlorophenyl cyanomethyl toluene, 4-amino-2-chloro-5-methylphenyl 4-chlorophenyl acetonitrile, 4-amino-2-chloro-alpha-4-chlorophenyl-5-methylbenzeneacetonitrile, 2-amino-4-chloro-5-4'-chlorophenyl cyanomethyl toluene, 2-amino-4-chloro-5-4?-chlorophenyl cyanomethyl toluene, 5-chloro-4-4-chloro-phenyl-cyano-methyl-2-methylanilin, 2-4-amino-2-chloro-5-methyl-phenyl-2-4-chlorophenyl acetonitrile, 2-4-amino-2-chloro-5-methylphenyl-2-4-chlorophenyl ethanenitrile, 4-amino-2-chloro-.alpha.-4-chlorophenyl-5-methylbenzeneacetonitrile
PubChem CID 2723752
IUPAC Name 2-(4-amino-2-chloro-5-methylphenyl)-2-(4-chlorophenyl)acetonitrile
SMILES CC1=CC(=C(C=C1N)Cl)C(C#N)C2=CC=C(C=C2)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 148°C to 153°C
Color Beige
Infrared Spectrum Authentic
Linear Formula ClC6H4CH(CN)C6H2(Cl)(CH3)NH2
Quantity 50 g
Formula Weight 291.17
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 4-Amino-2-chloro-α-(4-chlorophenyl)-5-methylbenzeneacetonitrile, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.