Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-amino-2-chloro-6,7-dimethoxyquinazoline | 23680-84-4 | 98.0% | 239.66 | C10H10ClN3O2 | 25 G
SDP

Catalog No. 5000209021
Encompass_Preferred

ACDQ (4-amino-2-chloro-6,7-dimethoxyquinazoline) is a small-molecule research chemical used as an inducer of adenylated nucleotide accumulation and in DNA/RNA synthesis-related studies. It is supplied as a white to off-white solid with high purity and standard cold-storage recommendations.

  • High purity: 98.0%.
  • Molecular formula C10H10ClN3O2 and molecular weight 239.66.
  • Appearance: white to off-white solid.
  • Typical storage recommendations include 4°C for the solid and -80°C (6 months) or -20°C (1 month) for solutions, protect from light.
  • Commonly used in DNA/RNA synthesis and nucleotide accumulation studies.
  • Available in small-quantity laboratory pack sizes, including 25 g.

Catalog No. 50-002-09021 Supplier Medchemexpress LLC Supplier No. 25GHYW087019
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACDQ (4-amino-2-chloro-6,7-dimethoxyquinazoline) is a small-molecule research chemical used as an inducer of adenylated nucleotide accumulation and in DNA/RNA synthesis-related studies. It is supplied as a white to off-white solid with high purity and standard cold-storage recommendations.

  • High purity: 98.0%.
  • Molecular formula C10H10ClN3O2 and molecular weight 239.66.
  • Appearance: white to off-white solid.
  • Typical storage recommendations include 4°C for the solid and -80°C (6 months) or -20°C (1 month) for solutions, protect from light.
  • Commonly used in DNA/RNA synthesis and nucleotide accumulation studies.
  • Available in small-quantity laboratory pack sizes, including 25 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.