Learn More
Medchemexpress LLC 4-amino-2-chlorobenzoic acid | 2457-76-3 | MFCD00007772 | 99.8% | 171.58 | C7H6ClNO2 | 100 G
SDP
Catalog No. 5000301931
Encompass_Preferred
4-Amino-2-chlorobenzoic acid (4ACB) is a biochemical reagent and chemical intermediate commonly used in research and synthesis. Supplied as a solid powder with high purity, it is suitable for analytical, synthetic, and assay applications and includes recommended storage guidelines to preserve stability.
  • High purity of approximately 99.8% for reliable experimental results.
  • Solid powder form for convenient storage and handling.
  • Suitable as a chemical intermediate and analytical reagent in synthesis workflows.
  • Recommended storage: powder at -20°C for long-term storage or 4°C for shorter term.
  • Available in multiple pack sizes to support different research scales.
Catalog No. 50-003-01931 Supplier Medchemexpress LLC Supplier No. HYW016925100G
4-Amino-2-chlorobenzoic acid (4ACB) is a biochemical reagent and chemical intermediate commonly used in research and synthesis. Supplied as a solid powder with high purity, it is suitable for analytical, synthetic, and assay applications and includes recommended storage guidelines to preserve stability.
  • High purity of approximately 99.8% for reliable experimental results.
  • Solid powder form for convenient storage and handling.
  • Suitable as a chemical intermediate and analytical reagent in synthesis workflows.
  • Recommended storage: powder at -20°C for long-term storage or 4°C for shorter term.
  • Available in multiple pack sizes to support different research scales.