Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-amino-2-chlorobenzoic acid | 2457-76-3 | MFCD00007772 | 99.8% | 171.58 g/mol | C7H6ClNO2 | 5 G
SDP

Catalog No. 5000301058
Encompass_Preferred

4-amino-2-chlorobenzoic acid is a substituted aminobenzoic acid used as a biochemical reagent and synthetic intermediate in organic chemistry and analytical research. It is intended for research use only and is commonly applied in synthesis, assay development, and other laboratory workflows.

  • CAS number: 2457-76-3.
  • Molecular formula: C7H6ClNO2.
  • Molecular weight: 171.58 g/mol.
  • Purity: 99.8%.
  • Appearance: off-white to light yellow solid.
  • Suited for use as a reagent and intermediate in laboratory synthesis.

Catalog No. 50-003-01058 Supplier Medchemexpress LLC Supplier No. HYW0169255G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-amino-2-chlorobenzoic acid is a substituted aminobenzoic acid used as a biochemical reagent and synthetic intermediate in organic chemistry and analytical research. It is intended for research use only and is commonly applied in synthesis, assay development, and other laboratory workflows.

  • CAS number: 2457-76-3.
  • Molecular formula: C7H6ClNO2.
  • Molecular weight: 171.58 g/mol.
  • Purity: 99.8%.
  • Appearance: off-white to light yellow solid.
  • Suited for use as a reagent and intermediate in laboratory synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.