Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-amino-2-chlorobenzoic acid | 2457-76-3 | MFCD00007772 | 99.8% | 171.58 g/mol | C7H6ClNO2 | 50 G
SDP

Catalog No. 5000301233
Encompass_Preferred

4-Amino-2-chlorobenzoic acid is a substituted benzoic acid (CAS 2457-76-3) commonly used as a building block and analytical reference in organic synthesis and biochemical research. Supplied as a powder with supporting analytical data, it is suitable for synthetic transformations, method development, and use as a reference standard.

  • High purity (HPLC 99.75%).
  • Molecular formula C7H6ClNO2.
  • Molecular weight 171.58 g/mol.
  • Available in multiple laboratory pack sizes.
  • Delivered with COA, SDS, and analytical data (HNMR, LCMS).
  • Store under recommended conditions to maintain stability.

Catalog No. 50-003-01233 Supplier Medchemexpress LLC Supplier No. HYW01692550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

4-Amino-2-chlorobenzoic acid is a substituted benzoic acid (CAS 2457-76-3) commonly used as a building block and analytical reference in organic synthesis and biochemical research. Supplied as a powder with supporting analytical data, it is suitable for synthetic transformations, method development, and use as a reference standard.

  • High purity (HPLC 99.75%).
  • Molecular formula C7H6ClNO2.
  • Molecular weight 171.58 g/mol.
  • Available in multiple laboratory pack sizes.
  • Delivered with COA, SDS, and analytical data (HNMR, LCMS).
  • Store under recommended conditions to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.