Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-amino-2-chlorobenzoic acid | 2457-76-3 | MFCD00007772 | 99.8% | 171.58 g/mol | C7H6ClNO2 | 500 G
SDP

Catalog No. 5000301057
Encompass_Preferred

4-Amino-2-chlorobenzoic acid is a research-grade biochemical reagent (CAS 2457-76-3) used as a synthetic building block and assay component. Supplied as an off-white to light yellow solid, it includes documented analytical data and is intended for research use only.

  • High purity: 99.8%.
  • Molecular weight 171.58 g/mol.
  • Chemical formula C7H6ClNO2.
  • Available in multiple pack sizes up to 500 g.
  • Solid form stable with recommended cold storage for long-term retention.
  • Certificate of analysis and safety data sheet available.

Catalog No. 50-003-01057 Supplier Medchemexpress LLC Supplier No. HYW016925500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Amino-2-chlorobenzoic acid is a research-grade biochemical reagent (CAS 2457-76-3) used as a synthetic building block and assay component. Supplied as an off-white to light yellow solid, it includes documented analytical data and is intended for research use only.

  • High purity: 99.8%.
  • Molecular weight 171.58 g/mol.
  • Chemical formula C7H6ClNO2.
  • Available in multiple pack sizes up to 500 g.
  • Solid form stable with recommended cold storage for long-term retention.
  • Certificate of analysis and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.