Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-chlorophenylboronic acid pinacol ester, 97%

Catalog No. AC444480010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444480010 1 g
AC444480050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC444480010 Supplier Thermo Scientific Chemicals Supplier No. 444480010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 877160-63-9 | C12H17BClNO2 | 253.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877160-63-9
Molecular Formula C12H17BClNO2
Molecular Weight (g/mol) 253.53
MDL Number MFCD09864184
InChI Key YKQXROMICPMQBZ-UHFFFAOYSA-N
Synonym 3-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-2-chlorophenylboronic acid, pinacol ester, 4-amino-2-chlorophenylboronic acid pinacol ester, 4-amino-2-chlorophenylboronic acid,pinacolester, 4-amino-2-chlorophenyl boronic acid pinacol ester, ykqxromicpmqbz-uhfffaoysa-n, 3-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl amine, 3-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-phenylamine, benzenamine, 3-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 44755206
IUPAC Name 3-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=C(Cl)C=C(N)C=C1

Specifications

Melting Point 123°C to 125°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 253.54
Percent Purity 97%
Chemical Name or Material 4-Amino-2-chlorophenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.