Learn More
Medchemexpress LLC 4-Amino-2-chloropyrimidine | 7461-50-9 | 99.8% | 129.55 | 25 G
SDP
Catalog No. 5000402895
Encompass_Preferred
4-Amino-2-chloropyrimidine is a pyrimidine derivative that acts as a competitive inhibitor for AChE and BChE. It also demonstrates high inhibitory activity on GST enzyme activity in vitro. This product is intended for research use only.
  • Competitive inhibitor for AChE (Ki = 0.18 μM) and BChE (Ki = 1.324 μM)
  • High inhibitory activity on GST enzyme activity (IC50 = 0.037 μM, Ki = 0.047 μM) in vitro
  • Intended for research use
Catalog No. 50-004-02895 Supplier Medchemexpress LLC Supplier No. 25GHY30139
4-Amino-2-chloropyrimidine is a pyrimidine derivative that acts as a competitive inhibitor for AChE and BChE. It also demonstrates high inhibitory activity on GST enzyme activity in vitro. This product is intended for research use only.
  • Competitive inhibitor for AChE (Ki = 0.18 μM) and BChE (Ki = 1.324 μM)
  • High inhibitory activity on GST enzyme activity (IC50 = 0.037 μM, Ki = 0.047 μM) in vitro
  • Intended for research use