Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3,5-dichloropyridine, 97%

Catalog No. AC367221000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC367221000 100 g
AC367220050 5 g
AC367220250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC367221000 Supplier Thermo Scientific Chemicals Supplier No. 367221000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 22889-78-7 | C5H4Cl2N2 | 163.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22889-78-7
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.00
MDL Number MFCD00125023
InChI Key ISIQAMHROGZHOV-UHFFFAOYSA-N
Synonym 4-amino-3,5-dichloropyridine, 3,5-dichloro-4-aminopyridine, 3,5-dichloro-4-pyridinamine, 4-pyridinamine, 3,5-dichloro, 3,5-dichloro-4-amino pyridine, 3,5-dichloro-4-pyridine amine, 3,5-dichloro-pyridin-4-ylamine, 3,5-dichloro-4-pyridylamine, 3,5-dichloropyridin-4-ylamine, 4-amino-3,5-dichloro pyridine
PubChem CID 89888
IUPAC Name 3,5-dichloropyridin-4-amine
SMILES NC1=C(Cl)C=NC=C1Cl

Specifications

Melting Point 158.0°C to 160.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 100 g
Formula Weight 163.01
Percent Purity 97%
Chemical Name or Material 4-Amino-3,5-dichloropyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Call a POISON CENTER or doctor/physician if you feel unwell. Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 245-304-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.