Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-amino-3-bromobenzoic acid | 6311-37-1 | 216.03 | 10 G
SDP

Catalog No. 5000446874
Encompass_Preferred

4-Amino-3-bromobenzoic acid is a biochemical reagent primarily intended for research use. It serves as a fundamental biochemical assay reagent. This solid, white to off-white compound has a molecular weight of 216.03 and boasts a purity of 99.59%, ensuring high quality for various laboratory applications.

  • Intended for research use
  • Solid, white to off-white appearance
  • Purity of 99.59%
  • Molecular weight: 216.03
  • Stable under recommended storage conditions
  • Suitable for diverse laboratory applications

Catalog No. 50-004-46874 Supplier Medchemexpress LLC Supplier No. HYW00847210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Amino-3-bromobenzoic acid is a biochemical reagent primarily intended for research use. It serves as a fundamental biochemical assay reagent. This solid, white to off-white compound has a molecular weight of 216.03 and boasts a purity of 99.59%, ensuring high quality for various laboratory applications.

  • Intended for research use
  • Solid, white to off-white appearance
  • Purity of 99.59%
  • Molecular weight: 216.03
  • Stable under recommended storage conditions
  • Suitable for diverse laboratory applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.