Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Amino-3-bromobenzoic acid | 6311-37-1 | 216.03 | 25 G
SDP

Catalog No. 5000448064
Encompass_Preferred

4-Amino-3-bromobenzoic acid is a biochemical reagent intended for research use only. This solid, white to off-white compound has a molecular weight of 216.03 and a purity of 99.59%.

  • Biochemical reagent for research use
  • Solid, white to off-white appearance
  • Molecular weight of 216.03
  • Purity of 99.59%
  • CAS number: 6311-37-1
  • Stable under recommended storage conditions
  • Store at room temperature for 3 years

Catalog No. 50-004-48064 Supplier Medchemexpress LLC Supplier No. 25GHYW008472
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Amino-3-bromobenzoic acid is a biochemical reagent intended for research use only. This solid, white to off-white compound has a molecular weight of 216.03 and a purity of 99.59%.

  • Biochemical reagent for research use
  • Solid, white to off-white appearance
  • Molecular weight of 216.03
  • Purity of 99.59%
  • CAS number: 6311-37-1
  • Stable under recommended storage conditions
  • Store at room temperature for 3 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.