Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Amino-3-bromobenzoic acid | 6311-37-1 | 216.03 | 5 G
SDP

Catalog No. 5000447486
Encompass_Preferred

4-Amino-3-bromobenzoic acid is a biochemical reagent intended for research use only.

  • For laboratory chemical use and substance manufacturing
  • Solid appearance
  • White to off-white color
  • Molecular formula: C7H6BrNO2
  • Melting point: 211-215°C
  • Boiling point: 368.5°C at 760 mmHg

Catalog No. 50-004-47486 Supplier Medchemexpress LLC Supplier No. HYW0084725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Amino-3-bromobenzoic acid is a biochemical reagent intended for research use only.

  • For laboratory chemical use and substance manufacturing
  • Solid appearance
  • White to off-white color
  • Molecular formula: C7H6BrNO2
  • Melting point: 211-215°C
  • Boiling point: 368.5°C at 760 mmHg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.