Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-bromobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. A15785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15785G 5 g
A15781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15785G Supplier TCI America Supplier No. A15785G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 57946-63-1
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.02
MDL Number MFCD00042150
InChI Key QKRJIXSZTKOFTD-UHFFFAOYSA-N
Synonym 2-bromo-4-trifluoromethyl aniline, 4-amino-3-bromobenzotrifluoride, 2-bromo-4-trifluoromethyl-phenylamine, benzenamine, 2-bromo-4-trifluoromethyl, 3-bromo-4-aminobenzotrifluoride, 4-amino-3-bromo benzotrifluoride, buttpark 83\07-26, timtec-bb sbb000789, buttpark 44\03-51, 2-bromo-4-trifluoromethyl phenylamine
PubChem CID 93899
IUPAC Name 2-bromo-4-(trifluoromethyl)aniline
SMILES NC1=CC=C(C=C1Br)C(F)(F)F

Specifications

Quantity 5 g
Formula Weight 240.02
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 4-Amino-3-bromobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.