Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-hydroxy-1-naphthalenesulfonic acid, For Spectrophotometric Det. of Si, ≥95.0%, MilliporeSigma™ Supelco™

Catalog No. 111012656 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2656 100 g
11-101-2655 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2656 Supplier MilliporeSigma Supelco Supplier No. 08751100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Amino-3-hydroxy-1-naphthalenesulfonic acid is an aromatic amino-sulfonic acid.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00004019
Synonym 1-Amino-2-naphthol-4-sulfonic acid

Specifications

Melting Point 290°C
CAS 116-63-2
Linear Formula H2 NC10H5(OH)SO3 H
Quantity 100 g
Formula Weight 239.25
Percent Purity ≥95.0% (CHN); ≥95.0%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.