Learn More
Medchemexpress LLC 4-Amino-3-hydroxynaphthalene-1-sulfonic acid | 116-63-2 | 98.0% | 239.25 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0134301G
Encompass_Preferred
4-Amino-3-hydroxynaphthalene-1-sulfonic acid is used as a fluorescent dye. It also forms polypyrrole (PPy) conducting film with pyrrole and is employed in electrochemistry.
  • Utilized as a fluorescent dye.
  • Forms polypyrrole (PPy) conducting film with pyrrole.
  • Utilized in electrochemistry.
  • For research use only.
Catalog No. 50-004-54393