Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-iodobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. A28731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A28731G Supplier TCI America Supplier No. A28731G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2122-63-6
Molecular Formula C7H6INO2
Molecular Weight (g/mol) 263.03
MDL Number MFCD01660550
InChI Key FPLDDZRJTOXFCB-UHFFFAOYSA-N
Synonym benzoic acid, 4-amino-3-iodo, 4-amino-3-iodobenzoicacid, 4-amino-3-iodo-benzoic acid, pubchem14046, rarechem al bo 1498, 4-carboxy-2-iodoaniline, 4-amino-3-iodo benzoic, wln: zr bi dvq, 4-amino-3-iodo-benzoicaci, 3-iodo-4-aminobenzoic acid
PubChem CID 16461
IUPAC Name 4-amino-3-iodobenzoic acid
SMILES NC1=CC=C(C=C1I)C(O)=O

Specifications

Melting Point 208°C
Quantity 1 g
Formula Weight 263.03
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-iodobenzoic Acid
RTECSNumber DG2802500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.