Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Amino-3-iodopyridine | 88511-27-7 | 220.01 | 25 G
SDP

Catalog No. 5000449548
Encompass_Preferred

4-Amino-3-iodopyridine is a chemical compound suitable for laboratory research and the manufacture of substances. This product is a solid that is stable under recommended storage conditions, which include keeping the container tightly sealed in a cool, well-ventilated area and protecting it from light.

  • Solid appearance
  • Stable under recommended storage conditions
  • Protect from light
  • Recommended storage at 4°C
  • Melting point of 99.1-99.4°C
  • Boiling point of 335.1±27.0 °C

Catalog No. 50-004-49548 Supplier Medchemexpress LLC Supplier No. 25GHYW002945
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Amino-3-iodopyridine is a chemical compound suitable for laboratory research and the manufacture of substances. This product is a solid that is stable under recommended storage conditions, which include keeping the container tightly sealed in a cool, well-ventilated area and protecting it from light.

  • Solid appearance
  • Stable under recommended storage conditions
  • Protect from light
  • Recommended storage at 4°C
  • Melting point of 99.1-99.4°C
  • Boiling point of 335.1±27.0 °C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.