Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Amino-3-methoxybenzoic acid | 2486-69-3 | C8H9NO3 | 10 G
SDP

Catalog No. 5000434573
Encompass_Preferred

4-Amino-3-methoxybenzoic acid is identified as a drug intermediate. This compound is suitable for constructing benzothiazole compounds, particularly those with anti-breast cancer activity.

  • Drug intermediate
  • Used to construct benzothiazole compounds
  • Exhibits anti-breast cancer activity
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 2 years, -20°C for 1 year
  • Soluble in DMSO at 25 mg/mL (149.56 mM)

Catalog No. 50-004-34573 Supplier Medchemexpress LLC Supplier No. HY7557710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Amino-3-methoxybenzoic acid is identified as a drug intermediate. This compound is suitable for constructing benzothiazole compounds, particularly those with anti-breast cancer activity.

  • Drug intermediate
  • Used to construct benzothiazole compounds
  • Exhibits anti-breast cancer activity
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 2 years, -20°C for 1 year
  • Soluble in DMSO at 25 mg/mL (149.56 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.