Learn More
Medchemexpress LLC 4-Amino-3-methoxybenzoic acid | 2486-69-3 | C8H9NO3 | 25 G
SDP
Catalog No. 5000358577
Encompass_Preferred
4-Amino-3-methoxybenzoic acid is a drug intermediate that can be used to construct benzothiazole compounds with anti-breast cancer activity.
  • Used as a drug intermediate
  • Can be used to construct benzothiazole compounds
  • Shows anti-breast cancer activity
Catalog No. 50-003-58577 Supplier Medchemexpress LLC Supplier No. 25GHY75577
4-Amino-3-methoxybenzoic acid is a drug intermediate that can be used to construct benzothiazole compounds with anti-breast cancer activity.
  • Used as a drug intermediate
  • Can be used to construct benzothiazole compounds
  • Shows anti-breast cancer activity