Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. A22831G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A22831G Supplier TCI America Supplier No. A22831G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A pyridines building block suitable for laboratory reagent use, with a minimum purity of 98.0%.

Chemical Identifiers

CAS 1990-90-5
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD01704431
InChI Key VGJLGPCXUGIXRQ-UHFFFAOYSA-N
Synonym 4-Amino-3-picoline
PubChem CID 74811
IUPAC Name 3-methylpyridin-4-amine
SMILES CC1=CN=CC=C1N

Specifications

Melting Point 108°C
Color White-Yellow
Quantity 1 g
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-methylpyridine
RTECSNumber TJ5140000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.