Learn More
Medchemexpress LLC 4-amino-3-phenylbutanoic acid hydrochloride | 3060-41-1 | 98.0% | 215.68 | C10H14ClNO2 | 5 G
SDP
Catalog No. 5000208791
Encompass_Preferred
Phenibut hydrochloride is the hydrochloride salt of β-phenyl-GABA, an orally active GABAB receptor agonist used as a research reagent with anxiolytic and cognition-enhancing effects. Supplied as a high-purity powder for laboratory research and pharmacology studies.
  • Orally active GABAB receptor agonist used in pharmacology research.
  • Anxiolytic and nootropic effects relevant to central nervous system studies.
  • High purity suitable for analytical and experimental use.
  • Molecular weight 215.68 g/mol and chemical formula C10H14ClNO2.
  • Available in multiple pack sizes including 5 g.
Catalog No. 50-002-08791 Supplier Medchemexpress LLC Supplier No. HYW0413335G
Phenibut hydrochloride is the hydrochloride salt of β-phenyl-GABA, an orally active GABAB receptor agonist used as a research reagent with anxiolytic and cognition-enhancing effects. Supplied as a high-purity powder for laboratory research and pharmacology studies.
  • Orally active GABAB receptor agonist used in pharmacology research.
  • Anxiolytic and nootropic effects relevant to central nervous system studies.
  • High purity suitable for analytical and experimental use.
  • Molecular weight 215.68 g/mol and chemical formula C10H14ClNO2.
  • Available in multiple pack sizes including 5 g.