Learn More
Medchemexpress LLC 4-amino-3-phenylbutanoic acid hydrochloride | 3060-41-1 | 98.0% | 215.68 g/mol | C10H14ClNO2 | 50 G
SDP
Catalog No. 5000209178
Encompass_Preferred
Phenibut hydrochloride is the hydrochloride salt of β-phenyl-GABA, a GABAB receptor agonist used as a research chemical to study GABAergic signaling. It is supplied as a solid powder with typical analytical purity around 98.0%.
  • GABAB receptor agonist for neuropharmacology research
  • High purity (98.0%) suitable for analytical use
  • Solid powder form, easy to weigh and dissolve
  • Commonly used in studies of anxiolytic and cognitive effects
  • Available in multiple pack sizes, including 50 g
Catalog No. 50-002-09178 Supplier Medchemexpress LLC Supplier No. HYW04133350G
Phenibut hydrochloride is the hydrochloride salt of β-phenyl-GABA, a GABAB receptor agonist used as a research chemical to study GABAergic signaling. It is supplied as a solid powder with typical analytical purity around 98.0%.
  • GABAB receptor agonist for neuropharmacology research
  • High purity (98.0%) suitable for analytical use
  • Solid powder form, easy to weigh and dissolve
  • Commonly used in studies of anxiolytic and cognitive effects
  • Available in multiple pack sizes, including 50 g