Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-amino-3-phenylbutanoic acid hydrochloride | 3060-41-1 | MFCD00075860 | ≥98.0% | 215.68 | C10H14ClNO2 | 10 G
SDP

Catalog No. 5000208789
Encompass_Preferred

Phenibut hydrochloride is the hydrochloride salt of β-phenyl-γ-aminobutyric acid and functions as a GABAB receptor agonist used in neuroscience research to study GABAergic signaling.

  • acts as a GABAB receptor agonist.
  • white to off-white solid powder appearance.
  • chemical formula C10H14ClNO2, molecular weight 215.68.
  • CAS number 3060-41-1.
  • purity ≥98.0% by stated analysis.
  • available in solid packaging, including 10 g sizes.

Catalog No. 50-002-08789 Supplier Medchemexpress LLC Supplier No. HYW04133310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenibut hydrochloride is the hydrochloride salt of β-phenyl-γ-aminobutyric acid and functions as a GABAB receptor agonist used in neuroscience research to study GABAergic signaling.

  • acts as a GABAB receptor agonist.
  • white to off-white solid powder appearance.
  • chemical formula C10H14ClNO2, molecular weight 215.68.
  • CAS number 3060-41-1.
  • purity ≥98.0% by stated analysis.
  • available in solid packaging, including 10 g sizes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.