Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-4'-dimethylaminoazobenzene 97.0+%, TCI America™
SDP

Catalog No. A16721G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16721G 1 g
A16725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16721G Supplier TCI America Supplier No. A16721G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 539-17-3
Molecular Formula C14H16N4
Molecular Weight (g/mol) 240.31
MDL Number MFCD00010204
InChI Key BVRIUXYMUSKBHG-UHFFFAOYSA-N
Synonym n,n-dimethyl-4,4'-azodianiline, 4'-amino-dab, c.i. disperse black 3, adab, acetile diazo black n, acetile diazo black r, 4-amino-4'-dimethylaminoazobenzene, supracet diazo black a, diazo nero microsetile g, microsetile diazo black g
PubChem CID 10872
IUPAC Name 4-[2-(4-aminophenyl)diazen-1-yl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(N)C=C1

Specifications

Melting Point 189°C
Color Red-Yellow
Quantity 1 g
Formula Weight 240.31
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-4′-dimethylaminoazobenzene
EINECSNumber (5)-4849
RTECSNumber BX5020000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.