Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-4'-nitrobiphenyl 96.0+%, TCI America™
SDP

Catalog No. A12511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12511G 1 g
A12515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12511G Supplier TCI America Supplier No. A12511G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1211-40-1
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.224
MDL Number MFCD00093527
InChI Key BOFVBIYTBGDQGY-UHFFFAOYSA-N
PubChem CID 14593
IUPAC Name 4-(4-nitrophenyl)aniline
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)[N+](=O)[O-])N

Specifications

Melting Point 204°C
Quantity 1 g
Synonym 4-Amino-4′C-nitrodiphenyl
Formula Weight 214.22
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-4′-nitrobiphenyl
RTECSNumber DV0350000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.