Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Amino-5-(2-phenylethyl)-4H-1,2,4-triazole-3-thiol

Catalog No. OTV0003331G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0003331G 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0003331G Supplier Sigma Aldrich Supplier No. OTV0003331G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-dodecyl-2-pyrrolidinone is an n-alkyl lactam and its reduction with lih3bnme2 to the corresponding amine has been reported
  • It is a potential melatonin-specific chemical penetration enhancer.1-dodecyl-2-pyrrolidinone was used in the preparation of few-layered flakes of molybdenum disulfide (mos2) via liquid phase exfoliation of bulk mos2 powder

Specifications

Molecular Formula C10H12N4S
Linear Formula C10H12N4S
Quantity 250 mL
MDL Number MFCD10689949
Molecular Weight (g/mol) 220.29
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.